Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNBP1 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB291465 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291465 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291465 Supplier Novus Biologicals Supplier No. NBP147260F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FNBP1 Polyclonal antibody specifically detects FNBP1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Microarray

Specifications

Antigen FNBP1
Applications Western Blot, Immunoprecipitation, Microarray
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation, Microarray
Formulation PBS
Gene Alias FBP17KIAA0554Formin-binding protein 17, formin binding protein 1, formin-binding protein 1, hFBP17, MGC126804
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 350 and 400 of human Formin-Binding Protein 17 using the numbering given in entry NP_055848.1 (GeneID 23048).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23048
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.