Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNBP1L Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176845 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176845 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176845 Supplier Novus Biologicals Supplier No. NBP176845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FNBP1L Polyclonal specifically detects FNBP1L in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen FNBP1L
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_001020119
Gene Alias C1orf39, chromosome 1 open reading frame 39, FLJ20275, formin binding protein 1-like, formin-binding protein 1-like, toca-1, TOCA1Transducer of Cdc42-dependent actin assembly protein 1, transducer of Cdc42-dependent actin assembly 1
Gene Symbols FNBP1L
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human TOCA-1. The immunogen is located within the last 50 amino acids of TOCA-1. Amino Acid Squence: GWTRARRQNGEEGYVPT
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54874
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.