Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNDC4 Antibody (OTI1B7), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB272151A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272151A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272151A48 Supplier Novus Biologicals Supplier No. NBP272151AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FNDC4 Monoclonal specifically detects FNDC4 in Human, Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen FNDC4
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI1B7
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias fibronectin type III domain containing 4, fibronectin type III domain-containing protein 4, Fibronectin type III repeat-containing protein 1, FRCP1FLJ22362
Gene Symbols FNDC4
Host Species Mouse
Immunogen Full length recombinant protein of human FNDC4 (NP_073734) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 64838.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.