Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNDC7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP238722 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP238722 0.1 mL
NB396697 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP238722 Supplier Novus Biologicals Supplier No. NBP238722
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FNDC7 Polyclonal specifically detects FNDC7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FNDC7
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q5VTL7
Gene Alias fibronectin type III domain containing 7, FLJ35838, MGC163293, RP11-293A10.2
Gene Symbols FNDC7
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: GINYTVTLKAISATGLTADCSYQSYFSGACCPLGVKLYRLGPNGIRIYWQASRGSANYSTDLYGSKGIFTCTPSAGLSFCDVTEIPCGDVYTVMV
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163479
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.