Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNDC8 Antibody (OTI1C10), HRP, Novus Biologicals™
SDP

Catalog No. NBP271966H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271966H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271966H Supplier Novus Biologicals Supplier No. NBP271966H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FNDC8 Monoclonal specifically detects FNDC8 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FNDC8
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1C10
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias DKFZp434H2215, fibronectin type III domain containing 8, fibronectin type III domain-containing protein 8
Gene Symbols FNDC8
Host Species Mouse
Immunogen Full length recombinant protein of human FNDC8 (NP_060029) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 54752.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.