Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNIP2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017252 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017252 Supplier Novus Biologicals Supplier No. NBP281941
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FNIP2 Polyclonal specifically detects FNIP2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen FNIP2
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA
Formulation PBS
Gene Alias FNIP1-like protein, FNIPL, folliculin interacting protein 2
Gene Symbols FNIP2
Host Species Rabbit
Immunogen FNIP2 antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human FNIP2. The immunogen is located within amino acids 100 - 150 of FNIP2. Amino Acid Squence: HHAKEQLPKYQYTRPASD
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57600
Test Specificity Multiple isoforms of FNIP2 are known to exist. This antibody is predicted to not cross-react with FNIP1.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.