Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoklR Rabbit anti-Bacteria, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP266850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266850 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266850 Supplier Novus Biologicals Supplier No. NBP266850
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fok1 Polyclonal specifically detects Fok1 in Bacteria samples. It is validated for Western Blot.

Specifications

Antigen FoklR
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000
Formulation PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias EC=3.1.21.4, Endonuclease FokI, FokI, Type II restriction enzyme FokI, Type IIS restriction enzyme FokI, Type-2 restriction enzyme FokI
Host Species Rabbit
Immunogen Synthetic peptide within Planomicrobium okeanokoites Fok1 aa 444-456. (SwissProt: P14870 Planomicrobium okeanokoites)
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.