Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Follistatin Antibody (17-2), PerCP, Novus Biologicals™
SDP

Catalog No. NB308558 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308558 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308558 Supplier Novus Biologicals Supplier No. NBP261941PCP

Mouse Monoclonal Antibody

Follistatin Monoclonal antibody specifically detects Follistatin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Follistatin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 17-2
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias follistatin, follistatin isoform FST317, FSActivin-binding protein
Host Species Mouse
Immunogen Recognizes human Follistatin isoform FS288, raised against recombinant human FS288
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10468
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.