Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Follistatin Antibody (FST/4281), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB378636 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378636 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378636 Supplier Novus Biologicals Supplier No. NBP308813UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Follistatin Monoclonal antibody specifically detects Follistatin in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Follistatin
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FST/4281
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias follistatin, follistatin isoform FST317, FSActivin-binding protein
Host Species Mouse
Immunogen Raised to human Follistatin (Fst) 288 and recognizes both human Fst 288 and 315 (Uniprot: P19883)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10468
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.