Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Follistatin Antibody (MM0296-9D28) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019165 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019165 0.025 mg
NBP212313 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019165 Supplier Novus Biologicals Supplier No. NBP2123130.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Follistatin Monoclonal specifically detects Follistatin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Follistatin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone MM0296-9D28
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:20 - 1:100 15ug/ml
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. P19883
Gene Alias follistatin, follistatin isoform FST317, FSActivin-binding protein
Gene Symbols FST
Host Species Mouse
Immunogen Human Follistatin
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10468
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.