Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Follistatin-like 1/FSTL1 Antibody (026), DyLight 755
SDP

Catalog No. NB108931 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB108931 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB108931 Supplier Novus Biologicals Supplier No. NBP289710IR

Rabbit Monoclonal Antibody

Follistatin-like 1/FSTL1 Monoclonal specifically detects Follistatin-like 1/FSTL1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Follistatin-like 1/FSTL1
Applications Immunofluorescence
Classification Monoclonal
Clone 26
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias FLJ50214, follistatin-like 1, Follistatin-like protein 1, follistatin-related protein 1, FRPFLJ52277, FSL1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Follistatin-like 1/FSTL1 (NP_009016.1; Met1-Ile308).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 11167
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.