Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Follistatin-related Gene Protein/FLRG/Fstl3 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB081341 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081341 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081341 Supplier Novus Biologicals Supplier No. NBP298213AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Follistatin-related Gene Protein/FLRG/Fstl3 Polyclonal specifically detects Follistatin-related Gene Protein/FLRG/Fstl3 in Mouse samples. It is validated for ELISA.

Specifications

Antigen Follistatin-related Gene Protein/FLRG/Fstl3
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias FLRGFSRP, follistatin-like 3 (secreted glycoprotein), Follistatin-like protein 3, Follistatin-related gene protein, follistatin-related protein 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Follistatin-related Gene Protein/FLRG/Fstl3 (NP_113557.1; Met1-Val256).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10272
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.