Learn More
Follistatin-related Gene Protein/FLRG/Fstl3 Antibody, DyLight 488, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Follistatin-related Gene Protein/FLRG/Fstl3 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | FLRGFSRP, follistatin-like 3 (secreted glycoprotein), Follistatin-like protein 3, Follistatin-related gene protein, follistatin-related protein 3 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Follistatin-related Gene Protein/FLRG/Fstl3 (NP_113557.1; Met1-Val256). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.