Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Follistatin-related Gene Protein/FLRG/Fstl3 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB321349 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB321349 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB321349 Supplier Novus Biologicals Supplier No. NBP298212MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Follistatin-related Gene Protein/FLRG/Fstl3 Polyclonal antibody specifically detects Follistatin-related Gene Protein/FLRG/Fstl3 in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Follistatin-related Gene Protein/FLRG/Fstl3
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias FLRGFSRP, follistatin-like 3 (secreted glycoprotein), Follistatin-like protein 3, Follistatin-related gene protein, follistatin-related protein 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse Follistatin-related Gene Protein/FLRG/Fstl3 (Accession#: NP_113557.1; Met 1-Val 256)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10272
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.