Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLR1 Antibody (548908) [DyLight 488], Novus Biologicals™
SDP

Catalog No. NB377753 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377753 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377753 Supplier Novus Biologicals Supplier No. FAB5646K
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FOLR1 Monoclonal antibody specifically detects FOLR1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Immunocytochemistry, CyTOF, Gene Knock-Out

Specifications

Antigen FOLR1
Applications Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Immunocytochemistry, CyTOF, Gene Knock-Out
Classification Monoclonal
Clone 548908
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Immunocytochemistry, CyTOF-ready, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias Adult folate-binding protein, FBP, folate binding protein, Folate receptor 1, folate receptor 1 (adult), folate receptor alpha, Folate receptor, adult, FOLR, FR-alpha, KB cells FBP, Ovarian tumor-associated antigen MOv18
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human FOLR1, Arg25-Met233, Accession # P15328
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2348
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.