Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLR1 Antibody (SF55 (BGN/1210/55)), Biotin, Novus Biologicals™
SDP

Catalog No. NB10062577B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062577B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062577B Supplier Novus Biologicals Supplier No. NB10062577B

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

FOLR1 Monoclonal specifically detects FOLR1 in Bovine samples. It is validated for ELISA.

Specifications

Antigen FOLR1
Applications ELISA
Classification Monoclonal
Clone SF55 (BGN/1210/55)
Conjugate Biotin
Dilution ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias Adult folate-binding protein, FBP, folate binding protein, Folate receptor 1, folate receptor 1 (adult), folate receptor alpha, Folate receptor, adult, FOLR, FR-alpha, KB cells FBP, Ovarian tumor-associated antigen MOv18
Gene Symbols FOLR1
Host Species Mouse
Immunogen Folate binding protein from bovine milk
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2348
Test Specificity NB100-62577 reacts with folate binding protein (FBP), a 30 kDa protein with a high affinity for folate and several reduced folic acid derivatives that mediates the intracelullar delivery of 5 methyltetrahydrofolate. Membrane bound and soluble forms of a high affinity FBP have been found in kidney, placenta, serum, milk, and several cell lines.
Target Species Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.