Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOLR1 Rabbit anti-Human, Mouse, Rat, Clone: 3J6W3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $504.00
Specifications
Antigen | FOLR1 |
---|---|
Clone | 3J6W3 |
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
FOLR1 Monoclonal antibody specifically detects FOLR1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
FOLR1 | |
Western Blot 1:500 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Adult folate-binding protein, FBP, folate binding protein, Folate receptor 1, folate receptor 1 (adult), folate receptor alpha, Folate receptor, adult, FOLR, FR-alpha, KB cells FBP, Ovarian tumor-associated antigen MOv18 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FOLR1 (NP_000793.1). MAQRMTTQLLLLLVWVAVVGEAQTRIAWARTELLNVCMNAKHHKEKPGPEDKLHEQCRPWRKNACCSTNTSQEAHKDVSYLYRFNWNHCGEMAPACKRHF | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
3J6W3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2348 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title