Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLR2 Antibody (001), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB109704 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB109704 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB109704 Supplier Novus Biologicals Supplier No. NBP289846AF405
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

FOLR2 Monoclonal specifically detects FOLR2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FOLR2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias BETA-HFR, FBP, FBP/PL-1, Folate receptor 2, folate receptor 2 (fetal), folate receptor beta, folate receptor, beta, Folate receptor, fetal/placental, folate-binding protein, fetal/placental, FR-BETA, FR-P3, Placental folate-binding protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human FOLR2 (P14207-1; Met1-His228).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2350
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.