Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLR2 Antibody (CL10), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB342859 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342859 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342859 Supplier Novus Biologicals Supplier No. NBP281079MFV450
Add to Cart
Add to Cart

Rat Monoclonal Antibody

FOLR2 Monoclonal antibody specifically detects FOLR2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen FOLR2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone CL10
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias BETA-HFR, FBP, FBP/PL-1, Folate receptor 2, folate receptor 2 (fetal), folate receptor beta, folate receptor, beta, Folate receptor, fetal/placental, folate-binding protein, fetal/placental, FR-BETA, FR-P3, Placental folate-binding protein
Host Species Rat
Immunogen FRbeta -transfected RBL2H3 cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2350
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.