Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLR4 Antibody (12A5), Novus Biologicals™
SDP

Catalog No. NBP226652SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226652SS 0.025 mg
NBP226652 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP226652SS Supplier Novus Biologicals Supplier No. NBP226652SS

Rat Monoclonal Antibody

FOLR4 Monoclonal specifically detects FOLR4 in Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular).

Specifications

Antigen FOLR4
Applications Flow Cytometry
Classification Monoclonal
Clone 12A5
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, Flow (Intracellular)
Gene Accession No. NP_075026, Q9EQF4
Gene Alias folate receptor 4 (delta) homolog (mouse), Folbp3, probable folate receptor delta
Gene Symbols FOLR4
Host Species Rat
Immunogen CD3 mAb-stimulated CD4/CD25 T cells
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390243
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.