Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOLR4 Antibody (TH6) - Azide Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP226657
Description
FOLR4 Monoclonal specifically detects FOLR4 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
FOLR4 | |
Monoclonal | |
Unconjugated | |
folate receptor 4 (delta) homolog (mouse), Folbp3, probable folate receptor delta | |
Rat | |
Protein G purified | |
RUO | |
390243 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2b κ |
Flow Cytometry, CyTOF | |
TH6 | |
Flow Cytometry 1ul/1 million cells, CyTOF-ready | |
IZUMO1R | |
CD3 mAb-stimulated CD4/25 T cells | |
0.05 mg | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction