Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOLR4 Antibody (TH6) - Azide Free, Novus Biologicals™

Rat Monoclonal Antibody
Specifications
Antigen | FOLR4 |
---|---|
Clone | TH6 |
Dilution | Flow Cytometry 1ul/1 million cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP226657
![]() |
Novus Biologicals
NBP226657 |
0.05 mg | N/A | N/A | N/A | ||||
Description
FOLR4 Monoclonal specifically detects FOLR4 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
FOLR4 | |
Flow Cytometry 1ul/1 million cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
folate receptor 4 (delta) homolog (mouse), Folbp3, probable folate receptor delta | |
IZUMO1R | |
IgG2b κ | |
Protein G purified |
TH6 | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Rat | |
Mouse | |
390243 | |
CD3 mAb-stimulated CD4/25 T cells | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title