Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLR4 Antibody (TH6), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP226657UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226657UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226657UV Supplier Novus Biologicals Supplier No. NBP226657UV
Add to Cart
Add to Cart

Rat Monoclonal Antibody

FOLR4 Monoclonal specifically detects FOLR4 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen FOLR4
Applications Flow Cytometry
Classification Monoclonal
Clone TH6
Conjugate DyLight 350
Dilution Flow Cytometry 1ul/1 million cells
Gene Alias folate receptor 4 (delta) homolog (mouse), Folbp3, probable folate receptor delta
Gene Symbols IZUMO1R
Host Species Rat
Immunogen CD3 mAb-stimulated CD4/25 T cells
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390243
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.