Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLR4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP19149620UL

 View more versions of this product

Catalog No. NBP19149620


Description

Description

FOLR4 Polyclonal specifically detects FOLR4 in Human samples. It is validated for Western Blot.
Specifications

Specifications

FOLR4
Polyclonal
Western Blot 1:1000
folate receptor 4 (delta) homolog (mouse), Folbp3, probable folate receptor delta
Rabbit
28 kDa
20 μL
Primary
Human
IgG
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
FOLR4
Synthetic peptide directed towards the middle region of human FOLR4. Peptide sequence TCKSNWRGGWDWSQGKNRCPKGAQCLPFSHYFPTPADLCEKTWSNSFKAS.
Affinity Purified
RUO
390243
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.