Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLR4 Rat anti-Mouse, DyLight™ 755, Clone: 12A5, Novus Biologicals™
SDP

Catalog No. NB001396 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001396 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001396 Supplier Novus Biologicals Supplier No. NBP226652IR0.1ML

Rat Monoclonal Antibody

FOLR4 Monoclonal specifically detects FOLR4 in Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular).

Specifications

Antigen FOLR4
Applications Flow Cytometry
Classification Monoclonal
Clone 12A5
Conjugate DyLight 755
Dilution Flow Cytometry, Flow (Intracellular)
Formulation 50 mM Sodium Borate
Gene Alias folate receptor 4 (delta) homolog (mouse), Folbp3, probable folate receptor delta
Gene Symbols IZUMO1R
Host Species Rat
Immunogen CD3 mAb-stimulated CD4/CD25 T cells
Quantity 0.1 mL
Gene ID (Entrez) 390243
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.