Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foot-and-mouth disease Antibody (2D2), Novus Biologicals™
SDP

Catalog No. NBP105452 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105452 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105452 Supplier Novus Biologicals Supplier No. NBP105452
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Foot-and-mouth disease Monoclonal antibody specifically detects Foot-and-mouth disease in Virus samples. It is validated for ELISA, Immunodiffusion.

Specifications

Antigen Foot-and-mouth disease
Applications ELISA, Immunodiffusion
Classification Monoclonal
Clone 2D2
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunodiffusion
Gene Alias fmdv
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with inactivated FMD virus.
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity specific for virulent serotype O1.
Target Species Virus
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.