Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Forkhead box I2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP120919 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120919 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP120919 Supplier Novus Biologicals Supplier No. NBP120919
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Forkhead box I2 Polyclonal specifically detects Forkhead box I2 in Mouse, Rat samples. It is validated for Peptide ELISA.

Specifications

Antigen Forkhead box I2
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_899016.1
Gene Alias FLJ46831, forkhead box I2, forkhead box protein I2, homolog of mouse Foxi2
Gene Symbols FOXI2
Host Species Goat
Immunogen Peptide with sequence C-GHLIYSRDGTE corresponding to C-Terminus according to Uniprot Mouse NP_899016.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399823
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.