Learn More
Specifications
Specifications
| Melting Point | 0°C |
| Color | Colorless |
| Boiling Point | 101°C |
| Assay Percent Range | 36.5 to 38.0 % |
| Packaging | PolyPac™ |
| Linear Formula | HCHO |
| Quantity | 20 L |
| Formula Weight | 30.03 |
| Percent Purity | 36.5 to 38.0% |
| Grade | Histological |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. NFPA Health:3 Flammability:2 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.