Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde Solution, 10% w/w in water, Fisher Chemical

Catalog No. sf9620 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
PolyPac™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SF96-20 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SF96-20

Chemical Identifiers

CAS 50-00-0 , 67-56-1 , 7732-18-5
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Melting Point 0°C
Color Colorless
Boiling Point 100°C
Assay Percent Range 10% w/w
Packaging PolyPac™
Quantity 20 L
Percent Purity 9.5 to 10.5%
Concentration or Composition (by Analyte or Components) 10% w/w
Physical Form Liquid
Chemical Name or Material Formaldehyde Solution, 10% w/w in 84.8 - 94.2% H2O
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Cancer hazard. Cannot be made non-poisonous. Harmful if absorbed through skin or if inhaled. Vapor harmful. Irritating to eyes, respiratory system and skin. May cause an allergic skin reaction. Possible risks of irreversible effects. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:2
Flammability:1
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.