Learn More
Formic acid, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Formic acid is principally used as a preservative antibacterial agent in livestock feed, in dying textiles, tanning leather and electroplating. In the poultry industry, it is sometimes added with feed to kill E. coli bacteria. It acts as a reducing agent for the catalytic reduction of nitrate in water.
Solubility
Miscible with water. Partially miscible with toluene, xylene and benzene.
Notes
Hygroscopic. Store closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
Specifications
Specifications
| Melting Point | 7°C to 9°C |
| Boiling Point | 100°C to 101°C |
| Linear Formula | HCOOH |
| UN Number | UN1779 |
| Beilstein | 1209246 |
| Merck Index | 144241 |
| Solubility Information | Miscible with water. Partially miscible with toluene,xylene and benzene. |
| Formula Weight | 46.03 |
| Percent Purity | 97% |
| Density | 1.22 |
| Show More |
Safety and Handling
| Hazard Category | H226-H302-H314-H331 |
| Hazard Statement | GHS H Statement H331-H314-H318-H226-H302 Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapour. Harmful if swallowed. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P312-P330-P331-P363-P372-P374-P380-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: FORMIC ACID |
| EINECSNumber | 200-579-1 |
| RTECSNumber | LQ4900000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.