Learn More
Thermo Scientific™ Formic Acid, LC-MS Grade PROMO
Description
Formic acid is a component found in reverse-phase mobile phases to provide protons for LC/MS analysis. The presence of a low concentration of formic acid in the mobile phase is also known to improve the peak shapes of the resulting separation. Unlike trifluoroacetic acid (TFA), formic acid is not an ion-pairing reagent, and it does not suppress MS ionization of polypeptides when used as a mobile phase component.
Ordering Alerts:
- X in the ordering table indicates that hazardous shipping charges apply.
Storage Requirements:
- Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Specifications
Specifications
| Standard Components | Formic acid |
| Standard Concentration | 98 to 100% |
| Item Name | Formic Acid |
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area |
| Form | Liquid |
| Reagent Type | Solvent (Pure) |
| Quantity | 10 x 1 mL |
| Formulation | 99% Formic Acid |
| Product Line | Pierce |
| For Use With | LC/MS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.