Learn More
Foss™ Cyclotec Sample Mill Accessories
Catalog No. TC10001989
Change view
Click to view available options
Pore Size:
0.03 in. (0.1 cm)
Catalog No. Pore Size
TC1000-1989 0.03 in. (0.1 cm)
1 options
Catalog No. TC1000-1989 Supplier Foss™ Supplier No. 10001989
For Cyclotec 1093 and Cyclotec 193, Cannot be used with the Cyclotec 293.
Specifications
Product Type Screen
For Use With (Equipment) Cyclotec™ sample mill
Pore Size 0.03 in. (0.1 cm)