Learn More
Eisco™ Fossiliferous Limestone Specimen
SCI_ED
Catalog No. S24728 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S24728 1 Ea.
S26607 10/Pk.
S24729 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S24728 Supplier Eisco™ Supplier No. ESNG0034
Add to Cart
Add to Cart
Fossiliferous Limestone Specimen for sedimentary rock identification and studying fossils within whole rocks.
  • Formed at bottom of marine environment through compaction of deceased organic matter in clay or silt cement
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Reacts strongly with hydrochloric acid, making it good for demonstrating effervescence
  • Made up of organic material, mostly CaCO3
Specifications
Product Type Sedimentary Rock
For Use With (Application) Studying fossils within whole rocks and sedimentary rock identification
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Fossiliferous Limestone