Learn More
Fotodyne™ FOTO/Analyst™ Investigator/FX Workstations with Manual Zoom, Side Lights
Catalog No. 1630F2DLNP
Click to view available options
:
Dual-wavelength UV dual-light transilluminator; w/PC, thermal printer; 8-bit, 1.4 Mpx camera
Dual-wavelength UV dual-light transilluminator; w/PC, thermal printer; 8-bit, 2.0 Mpx camera
Dual-wavelength UV dual-light transilluminator; w/PC; 8-bit, 1.4 Mpx camera
Dual-wavelength UV dual-light transilluminator; w/PC; 8-bit, 2.0 Mpx camera
Dual-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 1.4 Mpx camera
Dual-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 2.0 Mpx camera
Dual-wavelength UV transilluminator; w/PC; 8-bit, 1.4 Mpx camera
Dual-wavelength UV transilluminator; w/PC; 8-bit, 2.0 Mpx camera
Includes
8-48mm (6X) manual zoom lens, manual filter cassette, choice of CCD camera, choice of transilluminator, FOTO/FX darkroom with dual white-light and UV epi-illumination, image acquisition software, PCI frame grabber (with 0.4 megapixel camera) or firewire (with 1.4 and 2.0 megapixel cameras) card, ethidium bromide filter, Coomassie™ blue filter (dual-light workstations only), PC, optional thermal printer, all necessary cables and connectors
Large-format transilluminators provide the flexibility to capture images of many different sample types, including gels, microplates, fluorescent blots, colorimetric blots, TLC plates, and bacterial plates
  • System is designed to be easily upgraded to include a cooled CCD camera for low-light samples
  • Files can be archived in several formats to preserve data integrity for downstream analysis
  • Workstation can be networked to make files more accessible
  • Manual zoom lens allows adjustments to be made by rotating the on-lens aperature, zoom, and focus rings
FOTO/FX Benchtop Darkroom
  • Light-tight enclosure houses a CCD camera with zoom lens and filters
  • Flat black interior minimizes stray light
  • Gliding transilluminator platform and side-opening door with interlock provide convenient, safe operation of large-format transilluminators
  • Two interior visible epi-light panels allow easy focusing and even illumination of opaque samples
  • Dual UV epi-lights can be used to optimize sensitivity of samples stained with fluorescent dyes
  • Camera mounting system allows for easy upgrading to other cameras
  • Front-loading filter holder allows simple exchange of filters
FOTO/Flex Transillumators
  • Choose single-wavelength UV; single-wavelength UV, dual-light (UV plus white light); dual-wavelength UV; or dual-wavelength UV, dual-light models
  • UV transilluminators have 25 × 28cm UV glass; four 8 watt, 312nm (peak emission) bulbs; UV transparent protective cover; and preparative and analytical intensity settings
  • White-light transilluminators have a proprietary electroluminescent (EL) visible light panel that offers 15 times more uniform light illumination than traditional bulb-style transilluminators and continuously adjustable intensity control
Image Acquisition Software
  • Integration to 8.3 min. in 1/30 sec. increments
  • Live preview
  • Brightness and contrast
  • Image manipulation: invert, zoom, mirror, rotate, annotate (date, time, text, circle, arrow)
  • Image storage in choice of TIF, JPG, BMP, and PNG formats
Additional configurations are available. Contact your Fisher Scientific Customer Service Representative for details.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.