Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Four-Decade Resistance Box
SCI_ED

Catalog No. S05854 Shop All Eisco Products
Click to view available options
Class Size:
1 to 2 Lab Groups

The Four-Decade Resistance Box provides four-decade ranges from 0 to 11,110Ω.

  • Resolution of 1Ω over entire range.
  • Select decade by rotary switch.
  • Becomes an accurate potential divider by using tapping points across each range.
  • All resistance values are ±2% with rating of 0.25 w per range.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Provides four-decade ranges from 0 to 11,110 W
Includes 1 Apparatus
Class Size 1 to 2 Lab Groups
Product Type Four-decade Resistance Box
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.