Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOX2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15737320 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15737320 20 μL
NBP157373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15737320 Supplier Novus Biologicals Supplier No. NBP15737320UL

Rabbit Polyclonal Antibody

FOX2 Polyclonal specifically detects FOX2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FOX2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 4.0-8.0 ug/ml
Formulation PBS & 2% Sucrose. with No Preservative
Gene Alias dJ106I20.3, Fox-1 homolog B, fox-1 homologue, FOX2Fox-2, fxh, hexaribonucleotide binding protein 2, Hexaribonucleotide-binding protein 2, HRNBP2FOX-2, RBM9RTAHNRBP2, Repressor of tamoxifen transcriptional activity, RNA binding motif protein 9, RNA binding protein fox-1 homolog 2, RNA binding protein, fox-1 homolog (C. elegans) 2, RNA-binding motif protein 9, RNA-binding protein 9
Gene Symbols RBFOX2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to RBM9(RNA binding motif protein 9) The peptide sequence was selected from the middle region of RBM9. Peptide sequence TAAAAAAAAYSDGYGRVYTADPYHALAPAASYGVGAVASLYRGGYSRFAP.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23543
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.