Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXB1 Antibody (PCRP-FOXB1-1B7), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB178239 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB178239 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB178239 Supplier Novus Biologicals Supplier No. NBP314021AF350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FOXB1 Monoclonal antibody specifically detects FOXB1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen FOXB1
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-FOXB1-1B7
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias FKH5HFKH-5, forkhead box B1, forkhead box protein B1, Transcription factor FKH-5
Host Species Mouse
Immunogen Recombinant full-length human FOXB1protein (Uniprot: Q99853)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27023
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.