Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXB2 Antibody (PCRP-FOXB2-2B2), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB155329 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB155329 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB155329 Supplier Novus Biologicals Supplier No. NBP314125UV
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FOXB2 Monoclonal antibody specifically detects FOXB2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen FOXB2
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-FOXB2-2B2
Conjugate DyLight 350
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias bA159H20.4, forkhead box B2, forkhead box protein B2
Host Species Mouse
Immunogen Recombinant fragment (around aa1-200) of human FOXB2 protein (exact sequence is proprietary) (Uniprot: Q5VYV0)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 442425
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.