Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxC1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001268 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001268 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001268 Supplier Novus Biologicals Supplier No. NB1001268
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 12 publications

FoxC1 Polyclonal specifically detects FoxC1 in Human, Mouse, Zebrafish samples. It is validated for Chemotaxis, ChIP Assay, Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Peptide ELISA, Western Blot.

Specifications

Antigen FoxC1
Applications Chemotaxis, ChIP Assay, Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1.5 ug/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Peptide ELISA Detection limit 1:32000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockout Validated, Knockdown Validated
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q12948
Gene Alias ARA, FKHL7RIEG3, forkhead box C1, forkhead box protein C1, forkhead, drosophila, homolog-like 7, forkhead/winged helix-like transcription factor 7, forkhead-related activator 3, Forkhead-related protein FKHL7, Forkhead-related transcription factor 3, FREAC-3, FREAC3IRID1, IGDA, IHG1, myeloid factor-delta
Gene Symbols FOXC1
Host Species Goat
Immunogen FoxC1 Antibody is made to a peptide with sequence RTSGAFVYDCSKF corresponding to C-Terminus according to NP_001444.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2296
Target Species Human, Mouse, Zebrafish
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.