Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxC2 Antibody (OTI3E3), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP270752FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270752FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270752FR Supplier Novus Biologicals Supplier No. NBP270752FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FoxC2 Monoclonal specifically detects FoxC2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen FoxC2
Applications Western Blot
Classification Monoclonal
Clone OTI3E3
Conjugate DyLight 680
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias FKHL14LD, forkhead box C2 (MFH-1, mesenchyme forkhead 1), forkhead, Drosophila, homolog-like 14, Forkhead-related protein FKHL14, Mesenchyme fork head protein 1, mesenchyme forkhead 1, MFH-1, MFH-1 protein, MFH1forkhead box protein C2, Transcription factor FKH-14
Gene Symbols FOXC2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-280 of human FoxC2 (NP_005242) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2303.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.