Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXD1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP130959 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130959 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130959 Supplier Novus Biologicals Supplier No. NBP130959

Rabbit Polyclonal Antibody

FOXD1 Polyclonal antibody specifically detects FOXD1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen FOXD1
Applications Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q16676
Gene Alias FKHL8Forkhead, drosophila, homolog-like 8, forkhead box D1, forkhead-like 8, forkhead-related activator 4, Forkhead-related protein FKHL8, Forkhead-related transcription factor 4, FREAC-4, FREAC4forkhead box protein D1
Gene Symbols FOXD1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human FOXD1. The exact sequence is proprietary.
Molecular Weight of Antigen 46 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2297
Test Specificity Tested in human. Predicted cross-reactivity based on antigen sequence identity: Mouse (100%).
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.