Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxD3 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB319439 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB319439 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB319439 Supplier Novus Biologicals Supplier No. NBP298721MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FoxD3 Polyclonal antibody specifically detects FoxD3 in Human samples. It is validated for Western Blot

Specifications

Antigen FoxD3
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias AIS1, forkhead box D3, forkhead box protein D3, HFH2, HNF3/FH transcription factor genesis
Host Species Rabbit
Immunogen Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human FoxD3.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 27022
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.