Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxJ1/HFH4 Antibody (407003) [DyLight 488], Novus Biologicals™
SDP

Catalog No. NB371996 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371996 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371996 Supplier Novus Biologicals Supplier No. FAB3619K
Add to Cart
Add to Cart

Rat Monoclonal Antibody

FoxJ1/HFH4 Monoclonal specifically detects FoxJ1/HFH4 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen FoxJ1/HFH4
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 407003
Conjugate DyLight 488
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FKHL13forkhead transcription factor HFH-4, forkhead box J1, forkhead-like 13, Forkhead-related protein FKHL13, Hepatocyte nuclear factor 3 forkhead homolog 4, HFH4fork head homologue 4, HFH-4forkhead box protein J1, MGC35202
Host Species Rat
Immunogen E. coli-derived recombinant human FoxJ1, Lys306-Leu421, Accession # Q92949.3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2302
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.