Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXJ2 Antibody, Novus Biologicals™
SDP

Catalog No. NB035092 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB035092 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB035092 Supplier Novus Biologicals Supplier No. NBP287470
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FOXJ2 Polyclonal specifically detects FOXJ2 in Human samples. It is validated for Western Blot.

Specifications

Antigen FOXJ2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias FHXFork head homologous X, forkhead box J2, forkhead box protein J2, FOXJ2 forkhead factor
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of human FOXJ2
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55810
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.