Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXL2 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP270013R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270013R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270013R Supplier Novus Biologicals Supplier No. NBP270013R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FOXL2 Polyclonal specifically detects FOXL2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen FOXL2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3
Gene Symbols FOXL2
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human FOXL2 protein (between amino acids 350-376) [UniProt P58012]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 668.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.