Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXL2 Antibody, Novus Biologicals™
SDP

Catalog No. NB407717 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB407717 20 μg
NB1001277 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB407717 Supplier Novus Biologicals Supplier No. NB100127720UG
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 44 publications

FOXL2 Polyclonal specifically detects FOXL2 in Bovine, Chicken, Ferret, Human, Mouse samples. It is validated for Chemotaxis, ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry Whole-Mount, Peptide ELISA, Western Blot.

Specifications

Antigen FOXL2
Applications Chemotaxis, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (Whole-Mount), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 μg/mL, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 5 μg/mL, Immunohistochemistry-Frozen 1:10 - 1:500, Peptide ELISA Detection limit 1:128000, Immunohistochemistry (Whole-Mount) 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P58012
Gene Alias BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3
Gene Symbols FOXL2
Host Species Goat
Immunogen Peptide with sequence C-DSKTGALHSRLDL corresponding to C-Terminus according to NP_075555.1.
Purification Method Affinity Purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 668
Test Specificity Expected from sequence similiarity: Rat, Cow .
Target Species Bovine, Mouse, Chicken, Ferret, Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.