Learn More
FOXM1 Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | FOXM1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene | FOXM1 |
| Gene Accession No. | Q08050 |
| Gene Alias | AA408308; AW554517; BB238854; FKH L16; Fkh16; FKHL 16; FKHL16; forkhead box M1; forkhead box M1-like; Forkhead box protein M1; forkhead homolog 16; Forkhead like 16; Forkhead, drosophila, homolog-like 16; Forkhead-related protein FKHL16; FOXM 1; FOXM 1c; FOXM1; FOXM1B; Hepatocyte nuclear factor 3 forkhead homolog 11; HFH 11; HFH11; HFH-11; HFH-11B; HNF 3; HNF3; HNF-3; HNF-3/fork-head homolog 11; INS 1; INS1; INS-1; INS-1 winged helix; LOC100358944; M-phase phosphoprotein 2; MPHOSPH 2; MPHOSPH2; MPM 2; Mpm2; MPM-2 reactive phosphoprotein 2; MPP 2; MPP2; MPP-2; PIG 29; PIG29; TGT3; Transcription factor Trident; TRIDENT; WIN; Winged-helix factor from INS-1 cells; winged-helix transcription factor Trident |
| Gene Symbols | FOXM1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.