Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxO1/FKHR Antibody (83N7F8), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP231376X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231376X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP231376X Supplier Novus Biologicals Supplier No. NBP231376AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FoxO1/FKHR Monoclonal specifically detects FoxO1/FKHR in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen FoxO1/FKHR
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 83N7F8
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry 1:10-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Alias FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma
Gene Symbols FOXO1
Host Species Mouse
Immunogen FOXO1A
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2308
Test Specificity Human. Based upon immunogen sequence simlarity, this antibody is predicted to react with Mouse (92%), Rat (92%), Bovine/Cow (91%), Squirrel (91%) and Porcine/Pig (90%).
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.