Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxO1/FKHR Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP224857SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224857SS 0.025 mg
NBP224857 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224857SS Supplier Novus Biologicals Supplier No. NBP224857SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FoxO1/FKHR Polyclonal specifically detects FoxO1/FKHR in Human samples. It is validated for Western Blot.

Specifications

Antigen FoxO1/FKHR
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.05-0.50 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q12778
Gene Alias FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma
Gene Symbols FOXO1
Host Species Rabbit
Immunogen A portion of amino acids 250-300 of human FOXO1A protein was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2308
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat and bovine, 83% homologous in zebrafish and pufferfish, and 77% homologous in chicken.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.