Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxO1/FKHR Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
| Antigen | FoxO1/FKHR |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.05-0.50 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224857SS
![]() |
Novus Biologicals
NBP224857SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224857
![]() |
Novus Biologicals
NBP224857 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
FoxO1/FKHR Polyclonal specifically detects FoxO1/FKHR in Human samples. It is validated for Western Blot.Specifications
| FoxO1/FKHR | |
| Western Blot 0.05-0.50 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers | |
| PBS with 0.05% Sodium Azide | |
| FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma | |
| FOXO1 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat and bovine, 83% homologous in zebrafish and pufferfish, and 77% homologous in chicken. |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Q12778 | |
| 2308 | |
| A portion of amino acids 250-300 of human FOXO1A protein was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title